missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(4-methoxyphenyl)-3,4,9,10-perylenetetracarboxylic Diimide 97.0+%, TCI America™

Catalog No. B28921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28921G 1 g
B28925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28921G Supplier TCI America Supplier No. B28921G

Chemical Identifiers

CAS 6424-77-7
Molecular Formula C38H22N2O6
Molecular Weight (g/mol) 602.602
MDL Number MFCD00071978
InChI Key VZFVREBNFMQPSI-UHFFFAOYSA-N
Synonym Pigment Red 190
PubChem CID 80898
SMILES COC1=CC=C(C=C1)N2C(=O)C3=C4C(=CC=C5C4=C(C=C3)C6=C7C5=CC=C8C7=C(C=C6)C(=O)N(C8=O)C9=CC=C(C=C9)OC)C2=O

Specifications

Color Red-Yellow
Quantity 1 g
Formula Weight 602.60
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(4-methoxyphenyl)-3,4,9,10-perylenetetracarboxylic Diimide
EINECSNumber (5)-4072&(5)-4612
TSCA Yes