accessibility menu, dialog, popup

UserName

CH3ONOONOOOH3C

CAS RN 6424-77-7

IUPAC Name: 7,18-bis(4-methoxyphenyl)-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: pigment red 190
Molecular Weight (g/mol): 602.6
Molecular Formula: C38H22N2O6
InChi Key: VZFVREBNFMQPSI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1C(=O)C2=CC=C3C4=CC=C5C(=O)N(C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results