missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(4-chlorophenyl)-N,N'-diphenyl-1,4-phenylenediamine, TCI America™

Catalog No. B13361G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13361G Supplier TCI America Supplier No. B13361G

Chemical Identifiers

CAS 113703-66-5
Molecular Formula C30H22Cl2N2
Molecular Weight (g/mol) 481.42
MDL Number MFCD00060108
InChI Key SQYNUFZUOMNRHQ-UHFFFAOYSA-N
Synonym 4′C,4′C′C-Dichloro-N,N,N′C,N′C-tetraphenyl-1,4-phenylenediamine
PubChem CID 21851001
IUPAC Name 1-N,4-N-bis(4-chlorophenyl)-1-N,4-N-diphenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=CC=C3)C4=CC=C(C=C4)Cl)C5=CC=C(C=C5)Cl

Specifications

Melting Point 200°C
Quantity 1 g
Formula Weight 481.42
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(4-chlorophenyl)-N,N′-diphenyl-1,4-phenylenediamine