accessibility menu, dialog, popup

UserName

NNClCl

CAS RN 113703-66-5

IUPAC Name: N1,N4-bis(4-chlorophenyl)-N1,N4-diphenylbenzene-1,4-diamine
Synonyms: N1,N4-bis(4-chlorophenyl)-N1,N4-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol): 481.42
Molecular Formula: C30H22Cl2N2
InChi Key: SQYNUFZUOMNRHQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results