missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™

Catalog No. b4660200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4660200MG 200 mg
B46601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4660200MG Supplier TCI America Supplier No. B4660200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 850153-24-1
Molecular Formula C24H24BBr2NO2
Molecular Weight (g/mol) 529.079
InChI Key XVOQIMZHMJQJEC-UHFFFAOYSA-N
Synonym 2-[4-[Bis(4-bromophenyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-[Bis(4-bromophenyl)amino]phenylboronic Acid Pinacol Ester
PubChem CID 89535548
IUPAC Name N,N-bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br

Specifications

Melting Point 149°C
Color White
Quantity 200 mg
Formula Weight 529.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline