accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNBrBr

CAS RN 850153-24-1

IUPAC Name: N,N-bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N,N-bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 529.08
Molecular Formula: C24H24BBr2NO2
InChi Key: XVOQIMZHMJQJEC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results