missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline 98.0+%, TCI America™

Catalog No. p-7124102
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B55071G Supplier TCI America Supplier No. B55071G

Chemical Identifiers

CAS 663943-27-9
Molecular Formula C21H19Br2N
Molecular Weight (g/mol) 445.198
InChI Key XZCBYXUKPMELOQ-UHFFFAOYSA-N
PubChem CID 22930861
IUPAC Name N,N-bis(4-bromophenyl)-2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)C

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 1 g
Formula Weight 445.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline