accessibility menu, dialog, popup

UserName

CH3H3CNBrBrCH3

CAS RN 663943-27-9

IUPAC Name: N,N-bis(4-bromophenyl)-2,4,6-trimethylaniline
Synonyms: N,N-bis(4-bromophenyl)-2,4,6-trimethylaniline
Molecular Weight (g/mol): 445.2
Molecular Formula: C21H19Br2N
InChi Key: XZCBYXUKPMELOQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N(C2=CC=C(Br)C=C2)C2=CC=C(Br)C=C2)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results