missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine 98.0+%, TCI America™

Catalog No. B26195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26195G 5 g
B26191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26195G Supplier TCI America Supplier No. B26195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 351498-10-7
Molecular Formula C36H56N2O2
Molecular Weight (g/mol) 548.856
MDL Number MFCD06797077
InChI Key CLXKRTHAAROJRI-UHFFFAOYSA-N
PubChem CID 16735498
IUPAC Name 2,4-ditert-butyl-6-[[[3-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]-2,3-dimethylbutan-2-yl]amino]methylidene]cyclohexa-2,4-dien-1-one
SMILES CC(C)(C)C1=CC(=CNC(C)(C)C(C)(C)NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 548.86
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine