accessibility menu, dialog, popup

UserName

CH3H3CCH3HNH3CCH3H3CCH3NHOCH3H3CCH3CH3CH3H3COH3CH3CCH3(E/Z)(E/Z)

CAS RN 351498-10-7

IUPAC Name: 2,4-di-tert-butyl-6-{[(3-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-2,3-dimethylbutan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one
Synonyms: 2,4-di-tert-butyl-6-{[(3-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-2,3-dimethylbutan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 548.86
Molecular Formula: C36H56N2O2
InChi Key: CLXKRTHAAROJRI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CNC(C)(C)C(C)(C)NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results