missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-hydroxyethyl)oxamide 98.0+%, TCI America™

Catalog No. B17185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17185G 5 g
B171825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17185G Supplier TCI America Supplier No. B17185G

Chemical Identifiers

CAS 1871-89-2
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD00020559
InChI Key FPQJEXTVQZHURJ-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl oxamide, n1,n2-bis 2-hydroxyethyl oxalamide, nn'-bis 2-hydroxyethyl oxamide, unii-t92n2yr78x, ethanediamide, n,n'-bis 2-hydroxyethyl, n,n-bis 2-hydroxyethyl oxamide, n,n'-bis-2-hydroxyethyl-oxamide, ethanediamide, n1,n2-bis 2-hydroxyethyl, n,n'-bis 2-hydroxyethyl ethanediamide, n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide
PubChem CID 74638
IUPAC Name N,N'-bis(2-hydroxyethyl)oxamide
SMILES C(CO)NC(=O)C(=O)NCCO

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2-hydroxyethyl)oxamide