missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic Acid 98.0+%, TCI America™

Catalog No. p-7118738
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B090925G 25 g
B0909500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B090925G Supplier TCI America Supplier No. B090925G
Only null left

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid, bes, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, n,n-bis 2-hydroxyethyl taurine, n,n-bis hydroxyethyl-2-aminoethanesulfonic acid, bes buffering agent, taurine, n,n-bis 2-hydroxyethyl, 2-bis 2-hydroxyethyl amino-ethanesulfonicacid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Melting Point 155°C
Color White
Quantity 25 g
Formula Weight 213.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic Acid [Good′s buffer component for biological res
EINECSNumber (2)-3343
RTECSNumber KI7363500
TSCA Yes