missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-ethylhexyl)-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™

Catalog No. B4343200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4343200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4343200MG Supplier TCI America Supplier No. B4343200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82531-03-1
Molecular Formula C40H42N2O4
Molecular Weight (g/mol) 614.79
MDL Number MFCD21362484
InChI Key WEHOVFLILUDUKQ-UHFFFAOYNA-N
Synonym Plast Orange 8160
PubChem CID 87272327
IUPAC Name 7,18-bis(2-ethylhexyl)-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
SMILES CCCCC(CC)CN1C(=O)C2=CC=C3C4=CC=C5C(=O)N(CC(CC)CCCC)C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56

Specifications

Quantity 200 mg
Formula Weight 614.79
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2-ethylhexyl)-3,4,9,10-perylenetetracarboxylic Diimide