accessibility menu, dialog, popup

UserName

CAS RN 82531-03-1

IUPAC Name: 7,18-bis(2-ethylhexyl)-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: 7,18-bis(2-ethylhexyl)-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Weight (g/mol): 614.79
Molecular Formula: C40H42N2O4
InChi Key: WEHOVFLILUDUKQ-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C(=O)C2=CC=C3C4=CC=C5C(=O)N(CC(CC)CCCC)C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results