missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium N,N-Bis(carboxymethyl)-L-glutamate (ca. 40% in Water), TCI America™

Catalog No. B213525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B213525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B213525G Supplier TCI America Supplier No. B213525G

Chemical Identifiers

CAS 51981-21-6
Molecular Formula C9H9NNa4O8
Molecular Weight (g/mol) 351.129
MDL Number MFCD01862262
InChI Key UZVUJVFQFNHRSY-OUTKXMMCSA-J
Synonym N,N-Bis(carboxymethyl)-L-glutamic Acid Tetrasodium Salt
PubChem CID 44630158
IUPAC Name tetrasodium;(2S)-2-[bis(carboxylatomethyl)amino]pentanedioate
SMILES C(CC(=O)[O-])C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 351.13
Physical Form Liquid
Chemical Name or Material Tetrasodium N,N-Bis(carboxymethyl)-L-glutamate (ca. 40% in Water)