accessibility menu, dialog, popup

UserName

OOOONOOOONaNaNaNa

CAS RN 51981-21-6

IUPAC Name: tetrasodium 2-[bis(carboxylatomethyl)amino]pentanedioate
Synonyms: tetrasodium glutamate diacetate
Molecular Weight (g/mol): 351.13
Molecular Formula: C9H9NNa4O8
InChi Key: UZVUJVFQFNHRSY-UHFFFAOYNA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CCC(N(CC([O-])=O)CC([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results