missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 98.0+%, TCI America™
Supplier: TCI America B40595G
Specifications
| N,N-Bis(2-chloroethyl)-p-toluenesulfonamide | |
| 47°C | |
| C11H15Cl2NO2S | |
| 5 g | |
| PTVBBIMKLOMGSY-UHFFFAOYSA-N | |
| N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide | |
| 96039 | |
| ≥98.0% (HPLC,N) |
| 42137-88-2 | |
| White-Yellow | |
| MFCD00018944 | |
| n,n-bis 2-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-p-toluenesulphonamide, n,n-bis .beta.-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-toluenesulfonamide, n,n-bis 2'-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulphonamide, benzenesulfonamide, n,n-bis 2-chloroethyl-4-methyl, n,n-bis-2-chloro-ethyl-4-methyl-benzenesulfonamide | |
| CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl | |
| 296.206 | |
| 296.21 | |
| Crystalline Powder |
Chemical Identifiers
| 42137-88-2 | |
| 296.206 | |
| PTVBBIMKLOMGSY-UHFFFAOYSA-N | |
| 96039 | |
| CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl |
| C11H15Cl2NO2S | |
| MFCD00018944 | |
| n,n-bis 2-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-p-toluenesulphonamide, n,n-bis .beta.-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-toluenesulfonamide, n,n-bis 2'-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulphonamide, benzenesulfonamide, n,n-bis 2-chloroethyl-4-methyl, n,n-bis-2-chloro-ethyl-4-methyl-benzenesulfonamide | |
| N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide |
Safety and Handling
TSCA : No