missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. b405925g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B405925G 25 g
B40595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B405925G Supplier TCI America Supplier No. B405925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42137-88-2
Molecular Formula C11H15Cl2NO2S
Molecular Weight (g/mol) 296.206
MDL Number MFCD00018944
InChI Key PTVBBIMKLOMGSY-UHFFFAOYSA-N
Synonym n,n-bis 2-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-p-toluenesulphonamide, n,n-bis .beta.-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-toluenesulfonamide, n,n-bis 2'-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulphonamide, benzenesulfonamide, n,n-bis 2-chloroethyl-4-methyl, n,n-bis-2-chloro-ethyl-4-methyl-benzenesulfonamide
PubChem CID 96039
IUPAC Name N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 25 g
Formula Weight 296.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(2-chloroethyl)-p-toluenesulfonamide