missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,2-Tetramethyl-1,3-propanediamine 98.0+%, TCI America™

Catalog No. D1733500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1733500ML 500 mL
D173325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1733500ML Supplier TCI America Supplier No. D1733500ML
Only null left

Chemical Identifiers

CAS 53369-71-4
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.235
MDL Number MFCD00044018
InChI Key ULDIVZQLPBUHAG-UHFFFAOYSA-N
Synonym n,n,2,2-tetramethyl-1,3-propanediamine, 1,3-propanediamine, n,n,2,2-tetramethyl, n,n,2,2-tetramethylpropane-1,3-diamine, n,n-dimethylneopentanediamine, 3-amino-2,2-dimethylpropyl dimethylamine, n1,n1,2,2-tetramethylpropane-1,3-diamine, 1,3-propanediamine, n1,n1,2,2-tetramethyl, acmc-209l6k, ksc597o9f, uldivzqlpbuhag-uhfffaoysa
PubChem CID 104477
IUPAC Name N',N',2,2-tetramethylpropane-1,3-diamine
SMILES CC(C)(CN)CN(C)C

Specifications

Color Colorless
Boiling Point 153°C
Quantity 500 mL
UN Number 2734
Formula Weight 130.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,2,2-Tetramethyl-1,3-propanediamine