accessibility menu, dialog, popup

UserName

CAS RN 53369-71-4

CH3NCH3CH3H3CH2N

CAS RN 53369-71-4

IUPAC Name: (3-amino-2,2-dimethylpropyl)dimethylamine
Synonyms: (3-amino-2,2-dimethylpropyl)dimethylamine
Molecular Weight (g/mol): 130.24
Molecular Formula: C7H18N2
InChi Key: ULDIVZQLPBUHAG-UHFFFAOYSA-N
SMILES: CN(C)CC(C)(C)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results