missing translation for 'onlineSavingsMsg'
Learn More

N,N'-1,3-Phenylenedimaleimide 97.0+%, TCI America™

Catalog No. P097625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P097625G 25 g
P0976500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P097625G Supplier TCI America Supplier No. P097625G

Chemical Identifiers

CAS 3006-93-7
Molecular Formula C14H8N2O4
Molecular Weight (g/mol) 268.228
MDL Number MFCD00022569
InChI Key IPJGAEWUPXWFPL-UHFFFAOYSA-N
Synonym n,n'-1,3-phenylenedimaleimide, n,n'-m-phenylenedimaleimide, m-dimaleimidobenzene, m-phenylenedimaleimide, m-phenylenebismaleimide, 1,3-dimaleimidobenzene, hva 2, 1,3-bismaleimidobenzene, m-phdm, 1,3-phenylenebismaleimide
PubChem CID 18156
IUPAC Name 1-[3-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
SMILES C1=CC(=CC(=C1)N2C(=O)C=CC2=O)N3C(=O)C=CC3=O

Specifications

Melting Point 202°C
Quantity 25 g
UN Number 2811
Formula Weight 268.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-1,3-Phenylenedimaleimide
EINECSNumber (5)-3379&(9)-2054
RTECSNumber ON6125000
TSCA Yes