accessibility menu, dialog, popup

UserName

CAS RN 3006-93-7

OONNOO

CAS RN 3006-93-7

IUPAC Name: 1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[3-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 268.23
Molecular Formula: C14H8N2O4
InChi Key: IPJGAEWUPXWFPL-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC(=CC=C1)N1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results