missing translation for 'onlineSavingsMsg'
Learn More

N-Methylthiourea 98.0+%, TCI America™

Catalog No. p-7132269
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M061925G 25 g
M0619100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M061925G Supplier TCI America Supplier No. M061925G
Only null left

Chemical Identifiers

CAS 598-52-7
Molecular Formula C2H6N2S
Molecular Weight (g/mol) 90.144
MDL Number MFCD00004938
InChI Key KQJQICVXLJTWQD-UHFFFAOYSA-N
Synonym n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio
PubChem CID 2723704
IUPAC Name methylthiourea
SMILES CNC(=S)N

Specifications

Melting Point 120°C
Color White-Yellow
pH 7 (100g/L H2O soln. 20°C)
Quantity 25 g
UN Number 2811
Formula Weight 90.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylthiourea
RTECSNumber YT9000000
TSCA No