missing translation for 'onlineSavingsMsg'
Learn More

N-Methylthiourea, 97+%

Catalog No. AAA1043714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1043714 25 g
AAA1043722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1043714 Supplier Thermo Scientific Chemicals Supplier No. A1043714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 598-52-7 | C2H6N2S | 90.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-52-7
Molecular Formula C2H6N2S
Molecular Weight (g/mol) 90.144
MDL Number MFCD00004938
InChI Key KQJQICVXLJTWQD-UHFFFAOYSA-N
Synonym n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio
PubChem CID 2723704
IUPAC Name methylthiourea
SMILES CNC(=S)N

Specifications

Melting Point 119°C to 124°C
pH 7
Quantity 25 g
UN Number UN2811
Beilstein 506162
Formula Weight 90.15
Percent Purity ≥98%
Chemical Name or Material N-Methylthiourea
Hazard Category H300-H360
Hazard Statement GHS H Statement
H300-H361
Fatal if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-936-6
RTECSNumber YT9000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only