missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrolidone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. p-7111797
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
6002270 6 x 1 L Glass Bottle
6002274 4 x 4 L Glass Bottle
2 options

Catalog No. 6002270

Supplier: Honeywell Chemicals 3041L

Only null left

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Absorbance 0.5 AU max. at 300 nm, 0.1 AU max. at 325 nm, 0.03 AU max. at 350 nm, 0.01 AU max. at 400 nm
CAS Max % 100.0000%
Density 1.0259 g/cm3
Boiling Point 202°C
Flash Point 88°C
Packaging Glass Bottle
Quantity 6 x 1 L
Beilstein 106420
Formula Weight 99.15
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material N-Methylpyrrolidone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture