missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-2-carboxaldehyde, 98%

Catalog No. AC156930050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156930050 5 g Glass bottle
AC156930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156930050 Supplier Thermo Scientific Chemicals Supplier No. 156930050

CAS: 1192-58-1 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1192-58-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00003087
InChI Key OUKQTRFCDKSEPL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
PubChem CID 14504
IUPAC Name 1-methylpyrrole-2-carbaldehyde
SMILES CN1C=CC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.0100g/mL
Boiling Point 87°C to 90°C (22.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.562
Quantity 5 g
Beilstein 21, I, 279
Specific Gravity 1.01
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methylpyrrole-2-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 214-755-

RUO – Research Use Only