accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 1192-58-1

IUPAC Name: 1-methyl-1H-pyrrole-2-carbaldehyde
Synonyms: 1-methylpyrrole-2-carbaldehyde
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: OUKQTRFCDKSEPL-UHFFFAOYSA-N
SMILES: CN1C=CC=C1C=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)