missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyloctadecylamine 98.0+%, TCI America™
Supplier: TCI America M091225G
Specifications
| N-Methyloctadecylamine | |
| 44°C | |
| C19H41N | |
| 25 g | |
| SZEGKVHRCLBFKJ-UHFFFAOYSA-N | |
| N-methyloctadecan-1-amine | |
| 75539 | |
| ≥98.0% (GC,T) |
| 2439-55-6 | |
| White-Yellow | |
| MFCD00009008 | |
| N-Methylstearylamine | |
| CCCCCCCCCCCCCCCCCCNC | |
| 283.544 | |
| 283.54 | |
| Crystalline Powder |
Chemical Identifiers
| 2439-55-6 | |
| 283.544 | |
| SZEGKVHRCLBFKJ-UHFFFAOYSA-N | |
| 75539 | |
| CCCCCCCCCCCCCCCCCCNC |
| C19H41N | |
| MFCD00009008 | |
| N-Methylstearylamine | |
| N-methyloctadecan-1-amine |
Safety and Handling
RTECSNumber : RG4460000
TSCA : Yes