missing translation for 'onlineSavingsMsg'
Learn More

N-Methyloctadecylamine 98.0+%, TCI America™

Catalog No. p-7132438
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09125G 5 g
M091225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09125G Supplier TCI America Supplier No. M09125G

Chemical Identifiers

CAS 2439-55-6
Molecular Formula C19H41N
Molecular Weight (g/mol) 283.544
MDL Number MFCD00009008
InChI Key SZEGKVHRCLBFKJ-UHFFFAOYSA-N
Synonym N-Methylstearylamine
PubChem CID 75539
IUPAC Name N-methyloctadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCNC

Specifications

Melting Point 44°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyloctadecylamine
RTECSNumber RG4460000
TSCA Yes