missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylmaleimide 98.0+%, TCI America™
$246.05 - $2180.95
Chemical Identifiers
| CAS | 930-88-1 |
|---|---|
| Molecular Formula | C5H5NO2 |
| Molecular Weight (g/mol) | 111.1 |
| MDL Number | MFCD00005508 |
| InChI Key | SEEYREPSKCQBBF-UHFFFAOYSA-N |
| Synonym | n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione |
| PubChem CID | 70261 |
| IUPAC Name | 1-methylpyrrole-2,5-dione |
| SMILES | CN1C(=O)C=CC1=O |
Chemical Identifiers
| 930-88-1 | |
| 111.1 | |
| SEEYREPSKCQBBF-UHFFFAOYSA-N | |
| 70261 | |
| CN1C(=O)C=CC1=O |
| C5H5NO2 | |
| MFCD00005508 | |
| n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione | |
| 1-methylpyrrole-2,5-dione |
Specifications
| 930-88-1 | |
| White-Yellow | |
| MFCD00005508 | |
| 1759 | |
| SEEYREPSKCQBBF-UHFFFAOYSA-N | |
| 1-methylpyrrole-2,5-dione | |
| 70261 | |
| ≥98.0% (GC) | |
| N-Methylmaleimide |
| 97°C | |
| C5H5NO2 | |
| 25 g | |
| n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione | |
| CN1C(=O)C=CC1=O | |
| 111.1 | |
| 111.10 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : ON5600000
TSCA : No