missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleimide 98.0+%, TCI America™

Supplier:  TCI America M0807100G

 View more versions of this product

Catalog No. M0807100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Methylmaleimide
97°C
C5H5NO2
100 g
n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
CN1C(=O)C=CC1=O
111.1
111.10
Crystalline Powder
930-88-1
White-Yellow
MFCD00005508
1759
SEEYREPSKCQBBF-UHFFFAOYSA-N
1-methylpyrrole-2,5-dione
70261
≥98.0% (GC)

Chemical Identifiers

930-88-1
111.1
SEEYREPSKCQBBF-UHFFFAOYSA-N
70261
CN1C(=O)C=CC1=O
C5H5NO2
MFCD00005508
n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
1-methylpyrrole-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : ON5600000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions