missing translation for 'onlineSavingsMsg'
Learn More

N-Methylisobutylamine 98.0+%, TCI America™

Catalog No. p-7132303
Change view
Click to view available options
Quantity:
25 mL
100 mL
Catalog No. Quantity
M069125ML 25 mL
M0691100ML 100 mL
2 options

Catalog No. M069125ML

Supplier: TCI America M069125ML

Only null left

Chemical Identifiers

CAS 625-43-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015043
InChI Key QKYWADPCTHTJHQ-UHFFFAOYSA-N
Synonym n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin
PubChem CID 12249
IUPAC Name N,2-dimethylpropan-1-amine
SMILES CC(C)CNC

Specifications

Color Colorless
Boiling Point 78°C
Quantity 25 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylisobutylamine