missing translation for 'onlineSavingsMsg'
Learn More

N-Methyliminodiacetic Acid 98.0+%, TCI America™

Catalog No. p-7133018
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20905G 5 g
M209025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20905G Supplier TCI America Supplier No. M20905G

Chemical Identifiers

CAS 4408-64-4
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00004284
InChI Key XWSGEVNYFYKXCP-UHFFFAOYSA-N
Synonym n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
PubChem CID 20441
IUPAC Name 2-[carboxymethyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 5 g
Formula Weight 147.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyliminodiacetic Acid
EINECSNumber (2)-3726&(2)-3891
TSCA No