accessibility menu, dialog, popup

UserName

CH3NHOOOHO

CAS RN 4408-64-4

IUPAC Name: 2-[(carboxymethyl)(methyl)amino]acetic acid
Synonyms: [(carboxymethyl)(methyl)amino]acetic acid
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: XWSGEVNYFYKXCP-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)CC(O)=O
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)