missing translation for 'onlineSavingsMsg'
Learn More

N-Methyliminodiacetic acid, 99%

Catalog No. AAL0258906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0258906 5 g
AAL0258914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0258906 Supplier Thermo Scientific Chemicals Supplier No. L0258906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4408-64-4 | C5H9NO4 | 147.13 g/mol

N-Methyliminodiacetic acid is used to prepare bis-(1H-benzoimidazol-2-ylmethyl)-methyl-amine by condensation with benzene-1,2-diamine. It is involved in Suzuki cross-coupling with N-methyliminodiacetic acid (MIDA) boronates. It acts as a reagent utilized for protecting boronic acids and as a chelating agent for various metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyliminodiacetic acid is used to prepare bis-(1H-benzoimidazol-2-ylmethyl)-methyl-amine by condensation with benzene-1,2-diamine. It is involved in Suzuki cross-coupling with N-methyliminodiacetic acid (MIDA) boronates. It acts as a reagent utilized for protecting boronic acids and as a chelating agent for various metals.

Solubility
Soluble in water. Slightly soluble in dimethyl sulfoxide.

Notes
Incompatible with strong bases.

Chemical Identifiers

CAS 4408-64-4
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00004284
InChI Key XWSGEVNYFYKXCP-UHFFFAOYSA-N
Synonym n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
PubChem CID 20441
IUPAC Name 2-[carboxymethyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)CC(=O)O

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
Beilstein 1765449
Solubility Information Soluble in water. Slightly soluble in dimethyl sulfoxide.
Formula Weight 147.13
Percent Purity 99%
Chemical Name or Material N-Methyliminodiacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-557-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only