missing translation for 'onlineSavingsMsg'
Learn More

N-Methylhydroxylamine Hydrochloride 97.0+%, TCI America™

Supplier:  TCI America M080425G

 View more versions of this product

Catalog No. M080425G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Methylhydroxylamine Hydrochloride
86°C
CH6ClNO
25 g
RGZRSLKIOCHTSI-UHFFFAOYSA-N
hydrogen N-methylhydroxylamine chloride
77906
≥97.0% (T)
4229-44-1
White
MFCD00012597
n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
[H+].[Cl-].CNO
83.52
83.52
Crystalline Powder

Chemical Identifiers

4229-44-1
83.52
RGZRSLKIOCHTSI-UHFFFAOYSA-N
77906
[H+].[Cl-].CNO
CH6ClNO
MFCD00012597
n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
hydrogen N-methylhydroxylamine chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3528

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions