missing translation for 'onlineSavingsMsg'
Learn More

N-Methylhydroxylamine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7132370
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08045G 5 g
M080425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08045G Supplier TCI America Supplier No. M08045G
Only null left

Chemical Identifiers

CAS 4229-44-1
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012597
InChI Key RGZRSLKIOCHTSI-UHFFFAOYSA-N
Synonym n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
PubChem CID 77906
IUPAC Name hydrogen N-methylhydroxylamine chloride
SMILES [H+].[Cl-].CNO

Specifications

Melting Point 86°C
Color White
Quantity 5 g
Formula Weight 83.52
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylhydroxylamine Hydrochloride
EINECSNumber (2)-3528
TSCA Yes