missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylformanilide 99.0+%, TCI America™
Supplier: TCI America M0552100G
Specifications
| N-Methylformanilide | |
| Yellow | |
| C8H9NO | |
| 100 g | |
| JIKUXBYRTXDNIY-UHFFFAOYSA-N | |
| N-methyl-N-phenylformamide | |
| 66737 | |
| ≥99.0% (GC) |
| 93-61-8 | |
| 243°C | |
| MFCD00003283 | |
| n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide | |
| CN(C=O)C1=CC=CC=C1 | |
| 135.166 | |
| 135.17 | |
| Liquid |
Chemical Identifiers
| 93-61-8 | |
| 135.166 | |
| JIKUXBYRTXDNIY-UHFFFAOYSA-N | |
| 66737 | |
| CN(C=O)C1=CC=CC=C1 |
| C8H9NO | |
| MFCD00003283 | |
| n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide | |
| N-methyl-N-phenylformamide |
Safety and Handling
EINECSNumber : (3)-0111
TSCA : Yes