missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide 99.0+%, TCI America™

Catalog No. M0552500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0552500G 500 g
M055225G 25 g
M0552100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0552500G Supplier TCI America Supplier No. M0552500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-61-8
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00003283
InChI Key JIKUXBYRTXDNIY-UHFFFAOYSA-N
Synonym n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
PubChem CID 66737
IUPAC Name N-methyl-N-phenylformamide
SMILES CN(C=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 243°C
Quantity 500 g
Formula Weight 135.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylformanilide
EINECSNumber (3)-0111
TSCA Yes