missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide, 99%

CAS: 93-61-8 | C8H9NO | 135.166 g/mol

Supplier:  Thermo Scientific Chemicals A118290E

 View more versions of this product

Catalog No. AAA118290E


Only null left
Explore available promotions

Description

Description

N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

Solubility
Immisible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

N-Methylformanilide
9°C to 13°C
242°C to 244°C
C8H9NO
MFCD00003283
636496
Immisible with water.
CN(C=O)C1=CC=CC=C1
135.166
135.17
93-61-8
1.095
126°C (258°F)
1.561
2500 g
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
JIKUXBYRTXDNIY-UHFFFAOYSA-N
N-methyl-N-phenylformamide
66737
99%

Chemical Identifiers

93-61-8
135.166
JIKUXBYRTXDNIY-UHFFFAOYSA-N
66737
CN(C=O)C1=CC=CC=C1
C8H9NO
MFCD00003283
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
N-methyl-N-phenylformamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 202-262-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only