missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide, 99%

Catalog No. AAA1182936
Change view
Click to view available options
Quantity:
10,000 g
250 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1182936 500 g
AAA1182930 250 g
AAA118290E 2500 g
AAA118290C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1182936 Supplier Thermo Scientific Chemicals Supplier No. A1182936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-61-8 | C8H9NO | 135.166 g/mol

N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

Solubility
Immisible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 93-61-8
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00003283
InChI Key JIKUXBYRTXDNIY-UHFFFAOYSA-N
Synonym n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
PubChem CID 66737
IUPAC Name N-methyl-N-phenylformamide
SMILES CN(C=O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 13°C
Density 1.095
Boiling Point 242°C to 244°C
Flash Point 126°C (258°F)
Refractive Index 1.561
Quantity 500 g
Beilstein 636496
Solubility Information Immisible with water.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Methylformanilide
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-262-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only