missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformamide 99.0+%, TCI America™

Catalog No. p-7129471
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F005925G 25 g
F0059500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F005925G Supplier TCI America Supplier No. F005925G
Only null left

Chemical Identifiers

CAS 123-39-7
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00003280
InChI Key ATHHXGZTWNVVOU-UHFFFAOYSA-N
Synonym methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
PubChem CID 31254
ChEBI CHEBI:7438
IUPAC Name N-methylformamide
SMILES CNC=O

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 183°C
Quantity 25 g
Formula Weight 59.07
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylformamide
EINECSNumber (2)-0679
RTECSNumber LQ3000000
TSCA Yes