accessibility menu, dialog, popup

UserName

CAS RN 123-39-7

IUPAC Name: N-methylformamide
Synonyms: methylformamide NMF
Molecular Weight (g/mol): 59.07
Molecular Formula: C2H5NO
InChi Key: ATHHXGZTWNVVOU-UHFFFAOYSA-N
SMILES: CNC=O
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)