Learn More
N-Methylformamide, 99%
CAS: 123-39-7 | C2H5NO | 59.06 g/mol
Supplier: Thermo Scientific Chemicals 414330010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Methylformamide | |
| 123-39-7 | |
| 100.0 | |
| Colorless | |
| 199.0°C to 201.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4310 to 1.4330 | |
| MFCD00003280 | |
| 04, 58 | |
| 15, 6147 | |
| Solubility in water: miscible. Other solubilities: soluble in acetone and alcohol | |
| CNC=O | |
| 59.06 | |
| 1.9 mPa.s (20°C) | |
| 59.06 | |
| Liquid |
| 99+% | |
| 99.0 | |
| -3.2°C | |
| 1.0030g/mL | |
| 111°C | |
| 98.5% min. (GC) | |
| C2H5NO | |
| HCONHCH3 | |
| 1 kg | |
| 1.003 | |
| methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl | |
| ATHHXGZTWNVVOU-UHFFFAOYSA-N | |
| N-methylformamide | |
| 31254 | |
| CHEBI:7438 | |
| 99% |
Chemical Identifiers
| 123-39-7 | |
| 59.06 | |
| ATHHXGZTWNVVOU-UHFFFAOYSA-N | |
| 31254 | |
| N-methylformamide |
| C2H5NO | |
| MFCD00003280 | |
| methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl | |
| CHEBI:7438 | |
| CNC=O |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
May damage the unborn child.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
GHS Signal Word: Danger
EINECSNumber : 204-624-6
RTECSNumber : LQ3000000
TSCA : TSCA
RUO – Research Use Only