missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformamide, 99%

Catalog No. p-7043990
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0390818 50 g
AAL0390830 250 g
AAL039080B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0390818 Supplier Thermo Scientific Chemicals Supplier No. L0390818
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 123-39-7 | C2H5NO | 59.068 g/mol

N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.

Solubility
Miscible with water, acetone and ethanol. Immiscible with ether.

Notes
Incompatible with strong oxidizing agents, acids, bases and acid chlorides.

Chemical Identifiers

CAS 123-39-7
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00003280
InChI Key ATHHXGZTWNVVOU-UHFFFAOYSA-N
Synonym methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
PubChem CID 31254
ChEBI CHEBI:7438
IUPAC Name N-methylformamide
SMILES CNC=O

Specifications

Melting Point -4°C
Density 1.018
Boiling Point 181°C to 185°C
Flash Point 98°C (208°F)
Refractive Index 1.432
Linear Formula HCONHCH3
Quantity 50 g
Beilstein 1098352
Merck Index 14,6076
Solubility Information Miscible with water,acetone and ethanol. Immiscible with ether.
Formula Weight 59.07
Percent Purity 99%
Chemical Name or Material N-Methylformamide
Show More Show Less
Hazard Category H312-H360D
Hazard Statement GHS H Statement
H360-H312
May damage fertility or the unborn child.
Harmful in contact with skin.
Precautionary Statement P201-P202-P281-P302+P352-P308+P313-P312-P363-P501c
EINECSNumber 204-624-6
RTECSNumber LQ3000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only