missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 155560250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC155560250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

109-81-9
76.14
KFIGICHILYTCJF-UHFFFAOYSA-P
8014
C[NH2+]CC[NH3+]
C3H12N2
MFCD00008165
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
N'-methylethane-1,2-diamine

Specifications

N-Methylethylenediamine
94.0
0.8400g/mL
41°C
95%
C3H12N2
CH3NHCH2CH2NH2
MFCD00008165
0.84
KFIGICHILYTCJF-UHFFFAOYSA-P
N'-methylethane-1,2-diamine
8014
95%
109-81-9
100.0
114.0°C to 116.0°C
Authentic
Glass bottle
1.4390 to 1.4420
25g
04, I, 415
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
C[NH2+]CC[NH3+]
76.14
74.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-707-4

SDS
Documents

Documents

Promotions

Promotions