missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine, 95%, Thermo Scientific™

Catalog No. p-3779107
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155560050 5g Glass bottle
AC155560250 25g Glass bottle
AC155561000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155560050 Supplier Thermo Scientific Chemicals Supplier No. 155560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name N'-methylethane-1,2-diamine
SMILES C[NH2+]CC[NH3+]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.8400g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4390 to 1.4420
Linear Formula CH3NHCH2CH2NH2
Quantity 5g
Beilstein 04, I, 415
Specific Gravity 0.84
Formula Weight 74.13
Percent Purity 95%
Chemical Name or Material N-Methylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-707-4

RUO – Research Use Only