Learn More
N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126720010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Methyldiethanolamine | |
105-59-9 | |
100.0 | |
Colorless to Yellow | |
243°C | |
Authentic | |
Glass bottle | |
1.4675 to 1.4695 | |
MFCD00002848 | |
04, 284 | |
n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine | |
CRVGTESFCCXCTH-UHFFFAOYSA-N | |
2-[2-hydroxyethyl(methyl)amino]ethanol | |
7767 | |
119.16 | |
Liquid |
99+% | |
99.0 | |
-21°C | |
1.0380g/mL | |
138°C | |
99% min. (GC) | |
C5H13NO2 | |
(HOCH2CH2)2NCH3 | |
1 kg | |
1.038 | |
Solubility in water: miscible. Other solubilities: miscible with benzene,soluble in most organic solvents (alcohol,esters,aceton and chlorinated hydrocarbons),soluble in diethylether and aliphatic hydrocarbons | |
CN(CCO)CCO | |
119.16 | |
101 mPa.s (20°C) | |
99+% |
Chemical Identifiers
105-59-9 | |
119.16 | |
CRVGTESFCCXCTH-UHFFFAOYSA-N | |
7767 |
C5H13NO2 | |
MFCD00002848 | |
n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine | |
CN(CCO)CCO |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-312-7
RTECSNumber : KL7525000
TSCA : TSCA
RUO – Research Use Only