missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

Catalog No. p-162323
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126720010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC126720010 Supplier Thermo Scientific Chemicals Supplier No. 126720010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-59-9
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00002848
InChI Key CRVGTESFCCXCTH-UHFFFAOYSA-N
Synonym n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine
PubChem CID 7767
SMILES CN(CCO)CCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -21°C
Color Colorless to Yellow
Density 1.0380g/mL
Boiling Point 243°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4675 to 1.4695
Linear Formula (HOCH2CH2)2NCH3
Quantity 1 kg
Beilstein 04, 284
Specific Gravity 1.038
Solubility Information Solubility in water: miscible. Other solubilities: miscible with benzene,soluble in most organic solvents (alcohol,esters,aceton and chlorinated hydrocarbons),soluble in diethylether and aliphatic hydrocarbons
IUPAC Name 2-[2-hydroxyethyl(methyl)amino]ethanol
Viscosity 101 mPa.s (20°C)
Formula Weight 119.16
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Methyldiethanolamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-312-7
RTECSNumber KL7525000
TSCA TSCA

RUO – Research Use Only