missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcaprolactam, 96%

Catalog No. p-7023831
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1777614 25 g
AAA1777622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1777614 Supplier Thermo Scientific Chemicals Supplier No. A1777614
Only null left

CAS: 2556-73-2 | C7H13NO | 127.187 g/mol

N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.

Chemical Identifiers

CAS 2556-73-2
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00003263
InChI Key ZWXPDGCFMMFNRW-UHFFFAOYSA-N
Synonym n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
PubChem CID 17369
IUPAC Name 1-methylazepan-2-one
SMILES CN1CCCCCC1=O

Specifications

Density 0.991
Boiling Point 106°C to 108°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.484
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Fully miscible with water.
Formula Weight 127.18
Percent Purity 96%
Chemical Name or Material N-Methylcaprolactam
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 219-874-1
RTECSNumber CM4025000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only